missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575402
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The MRP family is represented by nine similar ABC transporters that have the ability to transport structurally diverse lipophilic anions and operate as chemical efflux pumps. MRP7 (multidrug resistance-associated protein 7, ATPbinding cassette sub-family C member 10) is a multi-pass membrane protein that belongs to the ABC transporter family (conjugate transporter subfamily). MRP7 is involved with the ATP-dependent transport of 17β-estradiol-D-17-β-glucuronide (E217βG). MRP7 is also probably involved in cellular detoxification through its lipophilic anion extrusion capabilities. MRP7 contains two ABC transmembrane type 1 domains and two ABC transporter domains. MRP7 likely has three isoforms. Isoform 2 is the most widely expressed, while isoform 1 is predominately expressed in the spleen.
Specifications
| MRP7 | |
| Polyclonal | |
| Unconjugated | |
| ABCC10 | |
| ABCC10; ATP binding cassette subfamily C member 10; ATP-binding cassette sub-family C member 10; ATP-binding cassette, subfamily C (CFTR/MRP), member 10; ATP-binding cassette, sub-family C (CFTR/MRP), member 10; EST182763; mFLJ00002; Mrp7; Multidrug resistance-associated protein 7; SIMRP7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 224814, 316231, 89845 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T3U5, Q8R4P9 | |
| ABCC10 | |
| Synthetic peptide corresponding to amino acids 834-887 of Human MRP7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction