missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP1 Antibody (IU2H10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB400-156APC
This item is not returnable.
View return policy
Description
MRP1 Monoclonal specifically detects MRP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| MRP1 | |
| Monoclonal | |
| APC | |
| P33527 | |
| ABCC1 | |
| Recombinant peptide (a.a. 1-33) of human MRP1. [UniProt# P33527]. | |
| 0.1 mL | |
| ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
| 4363 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| IU2H10 | |
| Flow Cytometry | |
| ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction