missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MROH7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584877
This item is not returnable.
View return policy
Description
Immunogen sequence: GIIILYTILE LQKRARDKEE TNKKELYESN KHFLGPYNPV SPCQNILRVI EEFGDFLGPQ QIKDLLLAAL EGLKGSSEAP GKDSREMMQL ASEVMLSSV Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 83%.
MROH7 is a protein coding gene.
Specifications
| MROH7 | |
| Polyclonal | |
| Unconjugated | |
| MROH7 | |
| C1orf175; Gm1027; HEAT repeat containing 8; HEAT repeat-containing protein 8; Heatr8; maestro heat like repeat family member 7; maestro heat-like repeat family member 7; maestro heat-like repeat-containing protein family member 7; MROH7; RGD1563440 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 374977 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68CQ1 | |
| MROH7 | |
| Recombinant protein corresponding to Human MROH7. Recombinant protein control fragment (Product #RP-108254). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction