missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554466
This item is not returnable.
View return policy
Description
Immunogen sequence: SCQLLLTILP RRQLPPGLES LNVSVAAGIL LHSICSQRKG FPTEGERRQL LQDPQEPSAR SEGLSMAQHP GLSS Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 58%.
MRM1 belongs to the RNA methyltransferase trmH family and probably methylates the ribose of guanosine G-2270 in the peptidyl transferase center of the mitochondrial large ribosomal RNA (21S).
Specifications
| MRM1 | |
| Polyclonal | |
| Unconjugated | |
| MRM1 | |
| 16S rRNA (guanosine(1145)-2'-O)-methyltransferase; 16S rRNA [Gm1145] 2'-O-methyltransferase; A530065E19Rik; CTB-75G16.3; ENSMUSG00000054212; mitochondrial large ribosomal RNA ribose methylase; mitochondrial rRNA methyltransferase 1; mitochondrial rRNA methyltransferase 1 homolog; mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae); Mrm1; RGD1566232; rRNA methyltransferase 1, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79922 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6IN84 | |
| MRM1 | |
| Recombinant protein corresponding to Human MRM1. Recombinant protein control fragment (Product #RP-93250). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction