missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRLC2 Recombinant Rabbit Monoclonal Antibody (24GB65)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557093
This item is not returnable.
View return policy
Description
MRLC2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
MRLC2 is involved in the the activity of nonmuscle myosin II is regulated by phosphorylation of a regulatory light chain. This phosphorylation results in higher MgATPase activity and the assembly of myosin II filaments.
Specifications
| MRLC2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MYL12B | |
| 1500001M02Rik; C77744; MLC-2; MLC20; MLC-2A; MLCB; MLC-B; Mrlc2; MRLC3; Mrlcb; Myl12b; MYLC2B; myosin light chain 12B; myosin light chain, regulatory B; myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain MRCL2; myosin regulatory light chain MRLC2; myosin regulatory light chain-B; myosin regulatory light polypeptide 9-like; myosin RLC-B; myosin, light chain 12B, regulatory; RLC; RLC-B; SHUJUN-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 103910, 50685, 67938 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB65 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14950, P18666, Q3THE2 | |
| MYL12B | |
| A synthesized peptide derived from human MYL12B (aa 150-172). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction