missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRLC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546308
This item is not returnable.
View return policy
Description
Peptide sequence: EKLNGTDPED VIRNAFACFD EEATGTIQED YLRELLTTMG DRFTDEEVDE Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 86%.
MRLC2 is involved in the the activity of nonmuscle myosin II is regulated by phosphorylation of a regulatory light chain. This phosphorylation results in higher MgATPase activity and the assembly of myosin II filaments.
Specifications
| MRLC2 | |
| Polyclonal | |
| Unconjugated | |
| MYL12B | |
| 1500001M02Rik; C77744; MLC-2; MLC20; MLC-2A; MLCB; MLC-B; Mrlc2; MRLC3; Mrlcb; Myl12b; MYLC2B; myosin light chain 12B; myosin light chain, regulatory B; myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain MRCL2; myosin regulatory light chain MRLC2; myosin regulatory light chain-B; myosin regulatory light polypeptide 9-like; myosin RLC-B; myosin, light chain 12B, regulatory; RLC; RLC-B; SHUJUN-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 103910 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O14950 | |
| MYL12B | |
| Synthetic peptide directed towards the C-terminal of human MYL12B (aa 92-141). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction