missing translation for 'onlineSavingsMsg'
Learn More

MRLC2 Antibody (1B22), Novus Biologicals™

Product Code. 18405019 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18405019 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18405019 Supplier Novus Biologicals Supplier No. H00103910M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRLC2 Monoclonal antibody specifically detects MRLC2 in Human samples. It is validated for ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRLC2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1B22
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_291024
Gene Alias MLC-2, MLC20, MLC-2A, MLC-B, MRLC2SHUJUN-1, MYLC2B, myosin regulatory light chain 12B, myosin regulatory light chain 2, Myosin regulatory light chain 20 kDa, Myosin regulatory light chain 2-B, smooth muscle isoform, Myosin regulatory light chain MRLC2, myosin, light chain 12B, regulatory
Host Species Mouse
Immunogen MRLC2 (NP_291024.1, 73 a.a. ∽ 172 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MMNEAPGPINFTMFLTMFGEKLNGTDPEDVIRNAFACFDEEATGTIQEDYLRELLTTMGDRFTDEEVDELYREAPIDKKGNFNYIEFTRILKHGAKDKDD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 103910
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.