missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19916-1-AP-150UL
This item is not returnable.
View return policy
Description
MGC3207 catalyzes the interconversion of methylthioribose-1-phosphate (MTR-1-P) into methylthioribulose-1-phosphate (MTRu-1-P). Independently from catalytic activity, MGC3207 promotes cell invasion in response to constitutive RhoA activation by promoting FAK tyrosine phosphorylation and stress fiber turnover.Specifications
| MRI1 | |
| Polyclonal | |
| Unconjugated | |
| MRI1 | |
| M1Pi, MRDI, MRI1, MTNA, MTR 1 P isomerase, Ypr118w | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288912, 67873, 84245 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5HZE4, Q9BV20, Q9CQT1 | |
| Mri1 | |
| MRI1 Fusion Protein Ag13809 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction