missing translation for 'onlineSavingsMsg'
Learn More

MRGX4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18362040 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362040 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362040 Supplier Novus Biologicals Supplier No. NBP2841710.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRGX4 Polyclonal specifically detects MRGX4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRGX4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias G protein-coupled receptor SNSR5, G protein-coupled receptor SNSR6, GPCR, MAS-related GPR, member X4, mas-related G-protein coupled receptor member X4, MGC129753, MGC129754, MRGX4G protein-coupled receptor MRGX4, Sensory neuron-specific G-protein coupled receptor 5/6, SNSR5, SNSR6
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of MRGX4
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 117196
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.