missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGX3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-44055
This item is not returnable.
View return policy
Description
MRGX3 Polyclonal specifically detects MRGX3 in Human samples. It is validated for Peptide ELISA.
Specifications
| MRGX3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| G protein-coupled receptor MRGX3, G protein-coupled receptor SNSR2, GPCR, mas-related GPCR member X3, MAS-related GPR, member X3, mas-related G-protein coupled receptor member X3, MRGX3G protein-coupled receptor SNSR1, Sensory neuron-specific G-protein coupled receptor 1/2, SNSR1, SNSR2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody may cross-react with human MRGPRX1 and X4 (GeneID: 259249, 117196; MW: 36.25kDa and 36.43kDa). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:4000 | |
| NP_473372.2 | |
| MRGPRX3 | |
| Peptide with sequence C-QLPQETLELSGSRL corresponding to internal region according to NP_473372.2. | |
| 0.1 mg | |
| GPCR | |
| 117195 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction