missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGX2 Antibody (477533), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4727U
This item is not returnable.
View return policy
Description
MRGX2 Monoclonal antibody specifically detects MRGX2 in Human samples. It is validated for Flow Cytometry, Flow Cytometry, CyTOF
Specifications
| MRGX2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 477533 | |
| Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGX2G protein-coupled receptor MRGX2 | |
| NS0 mouse myeloma cell line transfected with human MRGX2, Met1-Val330, Accession # Q96LB1 | |
| 0.1 mL | |
| GPCR | |
| 117194 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction