missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRGPRX4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533955
This item is not returnable.
View return policy
Description
MRGPRX4 Polyclonal Antibody for IHC (P)
Orphan receptor. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Potently activated by enkephalins.
Specifications
| MRGPRX4 | |
| Polyclonal | |
| Unconjugated | |
| MRGPRX4 | |
| G protein-coupled receptor MRGX4; G protein-coupled receptor SNSR5; G protein-coupled receptor SNSR6; GPCR; MAS related GPR family member X4; MAS-related GPR, member X4; mas-related G-protein coupled receptor member X4; MGC129753; MGC129754; MRGPRX4; MRGX4; sensory neuron-specific G-protein coupled receptor 5/6; SNSR5; SNSR6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117196 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q96LA9 | |
| MRGPRX4 | |
| Synthetic 12 amino acid peptide from N-terminal extracellular domain of human MRGPRX4 / MRGX4. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction