missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRGPRX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102263
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MRGX3.
Orphan receptor activated by a subset of RFamide-family neuropeptides such as FLRF-amide and FMRF-amide. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system. Its effect is mediated by G(q) and G(11) proteins. May regulate the function of nociceptive neurons by modulation of pain perception. Expressed in a subset of IB4-positive small diameter nociceptive dorsal root neurons.
Specifications
| MRGPRX3 | |
| Polyclonal | |
| Unconjugated | |
| MRGPRX3 | |
| adenine receptor; G protein-coupled receptor MRGX3; G protein-coupled receptor SNSR1; G protein-coupled receptor SNSR2; GPCR; G-protein coupled receptor; G-protein coupled receptor; rc 56.1.3; Hdrr1; HDRR2; MAS related GPR family member X3; Mas-related G protein-coupled receptor g1; Mas-related gene A; mas-related GPCR member X3; MAS-related GPR, member X3; mas-related G-protein coupled receptor member A; Mas-related G-protein coupled receptor member X3; MAS-related G-protein coupled receptor, member A; MGC108580; Mgrg1; Mrga; Mrga10; Mrgpra; MRGPRX3; MRGX3; nuclear receptor MrgA10 RF-amide G protein-coupled receptor; rc 56.1.3; sensory neuron-specific G-protein coupled receptor 1/2; SNSR1; SNSR2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117195 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96LB0 | |
| MRGPRX3 | |
| A synthesized peptide derived from human MRGPRX3(Accession Q96LB0), corresponding to amino acid residues P105-I155. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction