missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRGPRX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113198
This item is not returnable.
View return policy
Description
MRGPRX2 Polyclonal Antibody for ELISA, ICC/IF, Western Blot
Orphan receptor. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Cortistatin-14 seems to be a high potency ligand at this receptor. Cortistatin has several biological functions including roles in sleep regulation locomotor activity, and cortical function. In receptor-expressing cells, cortistatin-stimulated increases in intracellular Ca(2+) but had no effect on basal or forskolin-stimulated cAMP levels, suggesting that this receptor is G(q)-coupled. Has a limited expression profile, both peripheral and within the central nervous system, with highest levels in dorsal root ganglion.
Specifications
| MRGPRX2 | |
| Polyclonal | |
| Unconjugated | |
| MRGPRX2 | |
| G protein-coupled receptor MRGX2; G370024M05Rik; MAS related GPR family member X2; Mas-related G protein-coupled receptor G3; MAS-related GPR, member B10; MAS-related GPR, member X2; mas-related G-protein coupled receptor member B10; Mas-related G-protein coupled receptor member X2; MGRG3; MrgB10; Mrgprb10; Mrgprx2; MRGX2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 117194, 243978 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3UG50, Q96LB1 | |
| MRGPRX2 | |
| Recombinant Human Mas-related G-protein coupled receptor member X2 protein (286-330AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction