missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRGPRE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132771
This item is not returnable.
View return policy
Description
PA1-32771 detects GPCR MRGE from human samples.
Orphan receptor. May regulate nociceptor function and/or development, including the sensation or modulation of pain.
Specifications
| MRGPRE | |
| Polyclonal | |
| Unconjugated | |
| MRGPRE | |
| C130069N09Rik; Ebrt3; Evolutionary breakpoint transcript 3 protein; G protein-coupled receptor 167; G protein-coupled receptor MrgE; GPR167; G-protein coupled receptor 167; MAS related GPR family member E; mas-related G protein-coupled MRGE; Mas-related G protein-coupled receptor F; MAS-related gene E; MAS-related GPR, member E; mas-related G-protein coupled receptor member E; MAS-related G-protein coupled receptor, member E; MGC138408; Mgrf; MrgE; MRGPRE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116534 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q86SM8 | |
| MRGPRE | |
| Synthetic 17 amino acid peptide from C-terminus of human MRGE | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction