missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRGPRE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102103
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MRGRE.
Orphan receptor. May regulate nociceptor function and/or development, including the sensation or modulation of pain.
Specifications
| MRGPRE | |
| Polyclonal | |
| Unconjugated | |
| MRGPRE | |
| C130069N09Rik; Ebrt3; Evolutionary breakpoint transcript 3 protein; G protein-coupled receptor 167; G protein-coupled receptor MrgE; GPR167; G-protein coupled receptor 167; MAS related GPR family member E; mas-related G protein-coupled MRGE; Mas-related G protein-coupled receptor F; MAS-related gene E; MAS-related GPR, member E; mas-related G-protein coupled receptor member E; MAS-related G-protein coupled receptor, member E; MGC138408; Mgrf; MrgE; MRGPRE | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116534, 244238, 404660 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7TN40, Q86SM8, Q91ZB7 | |
| MRGPRE | |
| A synthesized peptide derived from human MRGPRE(Accession Q86SM8), corresponding to amino acid residues V180-G230. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction