missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRG15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA586490
This item is not returnable.
View return policy
Description
MRG15 ia a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair.
Specifications
| MRG15 | |
| Polyclonal | |
| Unconjugated | |
| MORF4L1 | |
| Mortality factor 4-like protein 1; MORF-related gene 15 protein; Protein MSL3-1; Transcription factor-like protein MRG15; MORF4L1; MRG15; FWP006; HSPC008; HSPC061; PP368 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10933, 21761, 300891 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.2 | |
| P60762, Q6AYU1, Q9UBU8 | |
| Morf4l1 | |
| Synthetic peptide, corresponding to amino acids 23-75 of Human MRG15. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction