missing translation for 'onlineSavingsMsg'
Learn More

MRG15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen MRG15
Dilution Western Blot 1:500 - 1:2000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30229449
View Documents
Novus Biologicals
NBP3-38377-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30227059
View Documents
Novus Biologicals
NBP3-38377-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MRG15 Polyclonal antibody specifically detects MRG15 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

MRG15
ELISA, Western Blot
Unconjugated
Rabbit
DNA replication Transcription Translation and Splicing
PBS (pH 7.3), 50% glycerol
10933
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA
Polyclonal
Purified
RUO
Human, Mouse, Rat
Eaf3, Esa1p-associated factor 3 homolog, HsT17725, MEAF3, MGC10631, MORF-related gene on chromosome 15, MORFRG15, mortality factor 4 like 1, mortality factor 4-like protein 1, MRG15MORF-related gene 15 protein, Protein MSL3-1, S863-6, Transcription factor-like protein MRG15
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human MRG15 (NP_006782.1).,, Sequence:, MAPKQDPKPKFQEGERVLCFHGPLLYEAKCVKVAIKDKQVKYFIHYSGWNKNWDEWVPESRVLKYVDTNLQKQRELQKANQEQYAEGKMRGAAPGKKTSG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.