missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRFAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576589
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes an intracellular protein that interacts with members of the MORF4/MRG (mortality factor on chromosome 4/MORF4 related gene) family and the tumor suppressor Rb (retinoblastoma protein.) The protein may play a role in senescence, cell growth and immortalization. Alternative splicing results in multiple transcript variants.
Specifications
| MRFAP1 | |
| Polyclonal | |
| Unconjugated | |
| MRFAP1 | |
| 9130413I22Rik; AL022827; MNCb-1039; Mof4 family associated protein 1; Morf4 family associated protein 1; Morf4 family associated protein 1 like 1; Morf4 family associated protein 1-like 1; MORF4 family-associated protein 1; MORF4 family-associated protein 1-like 1; MRFAP1; MRFAP1L1; PAM14; PGR1; PP784; protein associated with MRG of 14 kDa; protein associated with MRG, 14 kDa; Protein PGR1; T-cell activation protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y605 | |
| MRFAP1 | |
| Recombinant fusion protein of human MRFAP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction