missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRFAP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94813-0.02ml
This item is not returnable.
View return policy
Description
MRFAP1 Polyclonal antibody specifically detects MRFAP1 in Human samples. It is validated for Western Blot
Specifications
| MRFAP1 | |
| Polyclonal | |
| Western Blot 1:200-1:1000 | |
| Mof4 family associated protein 1, Morf4 family associated protein 1, PAM14MORF4 family-associated protein 1, PGR1protein associated with MRG, 14 kDa, Protein associated with MRG of 14 kDa, Protein PGR1, T-cell activation protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human MRFAP1 (NP_150638.1). MRPLDIVELAEPEEVEVLEPEEDFEQFLLPVINEMREDIASLTREHGRAYLRNRSKLWEMDNMLIQIKTQVEASEESALNHLQNPGDAAEGRAAKRCEKAEEKAKEIAKMAEMLVELVRRIEKSESS | |
| 0.02 mL | |
| Cell Biology | |
| 93621 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion