missing translation for 'onlineSavingsMsg'
Learn More

MRE3008F20, MedChemExpress

Product Code. 16466527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16466527 Supplier Medchem Express Supplier No. HY103178
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MRE3008F20 is a highly potent and selective antagonist of adenosine A3 receptor (AA3R), inhibiting agonist-induced cAMP elevation in resting T lymphocytes with an IC50 of 5 nM.

MRE3008F20 is a highly potent and selective antagonist of adenosine A3 receptor (AA3R), inhibiting agonist-induced cAMP elevation in resting T lymphocytes with an IC50 of 5 nM.

Specifications

Chemical Name or Material MRE3008F20
CAS 252979-43-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H20N8O3
Quantity 5 mg
SMILES O=C(NC1=CC=C(OC)C=C1)NC2=NC3=NN(CCC)C=C3C4=NC(C5=CC=CO5)=NN24
Molecular Weight (g/mol) 432.44
Formula Weight 432.44
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.