missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mre11-Rad50-NBS1 Complex Antibody Pack

Product Code. 18750694 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18750694 Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18750694 Supplier Novus Biologicals™ Supplier No. NB100914

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • 1.NB100-142: Mre11 Antibody
  • 2.NB100-143: Nbs1 Antibody
  • 3.NB100-154: Rad50 Antibody

For Research applications

TRUSTED_SUSTAINABILITY

Specifications

Applications ELISA, Immunohistochemistry
Conjugate Unconjugated
Gene Alias ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11MRE11B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.