missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mre11 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
£308.00
Specifications
| Antigen | Mre11 |
|---|---|
| Dilution | Western Blot, Immunoprecipitation |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 646 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18517943
|
Novus Biologicals
NBP2-99084JF646 |
0.1 ml | |||||||
Description
Mre11 Polyclonal antibody specifically detects Mre11 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Mre11 | |
| Western Blot, Immunoprecipitation | |
| Janelia Fluor 646 | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase | |
| ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Mre11. | |
| Primary |
| Western Blot, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| 4361 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title