missing translation for 'onlineSavingsMsg'
Learn More

Mre11 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18369162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369162 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369162 Supplier Novus Biologicals Supplier No. NBP299084AF700

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mre11 Polyclonal specifically detects Mre11 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mre11
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B
Host Species Rabbit
Immunogen Mre11 Antibody was produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Mre11.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 4361
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.