missing translation for 'onlineSavingsMsg'
Learn More

MRE-269, MedChemExpress

Product Code. 16490688
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490688

Brand: Medchem Express HY79593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MRE-269 is an active metabolite of selexipag, and acts as a selective IP receptor agonist.

MRE-269 is an active metabolite of selexipag, and acts as a selective IP receptor agonist.

Specifications

Chemical Name or Material MRE-269
Physical Form Solid
CAS 475085-57-5
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C25H29N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym ACT-333679
Solubility Information DMSO : 100 mg/mL (238.37 mM; Need ultrasonic)
SMILES OC(COCCCCN(C(C)C)C1=NC(C2=CC=CC=C2)=C(C3=CC=CC=C3)N=C1)=O
Molecular Weight (g/mol) 419.52
Formula Weight 419.52
Percent Purity 98.43%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.