missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRCL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86713
This item is not returnable.
View return policy
Description
MRCL3 Polyclonal specifically detects MRCL3 in Human samples. It is validated for Western Blot.
Specifications
| MRCL3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10627 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| MLCBMyosin RLC, MRCL3, MRLC3MLC-2B, MYL2B, myosin regulatory light chain 12A, Myosin regulatory light chain 2, nonsarcomeric, myosin regulatory light chain 3, myosin regulatory light chain MRCL3, Myosin regulatory light chain MRLC3, myosin, light chain 12A, regulatory, non-sarcomeric, myosin, light polypeptide, regulatory, non-sarcomeric (20kD), RLC | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of human MRCL3 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction