Learn More
Eurofins DiscoverX MRCKβ Protein, Active
Description
CDC42BPB (CDC42 binding protein kinase beta) is another member of the rho family and triggers the formation of actin-based structure found at lamellipodia and filopodia. CDC42BPB plays an important role in determining mammalian cell morphology for the coordinated control of cell motility by members of the rho family of GTPases.
GenBank NM_006035
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 55.5 kDa |
| Quantity | 10 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥65% |
| Protein | MRCKβ, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.