missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRCK beta Polyclonal Antibody

Description
This antibody detects endogenous protein at a molecular weight of 194 kDa. Purity is >95% by SDS-PAGE.
CDC42BPB is a serine/threonine kinases which functions in actin cytoskeleton assembly and organization. Myotonic dystrophy kinaserelated Cdc42-binding (DMPK-like) kinases-alpha and beta (MRCK-alpha, beta) contain a cysteine-rich motif and a putative pleckstrin homology domain. MRCKs can phosphorylate nonmuscle Myosin light chain and influences Actin-Myosin contractility. MRCK-alpha can phosphorylate and activate LIM kinases downstream of Cdc42, which leads to inactivation of ADF/Cofilin and to Actin cytoskeletal reorganization. MRCK-alpha can also influence neurite outgrowth promoted by Cdc42 and Rac.
Specifications
Specifications
| Antigen | MRCK beta |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CDC42BPB |
| Gene Accession No. | Q7TT49, Q7TT50, Q9Y5S2 |
| Gene Alias | CDC42 binding protein kinase beta; CDC42 binding protein kinase beta (DMPK-like); CDC42-binding protein kinase beta; CDC42-binding protein kinase beta (DMPK-like); CDC42BPB; cdc42bpb {ECO:0000312; CDC42BP-beta; DMPK-like beta; EMBL:AAP34403.1}; Kiaa1124; MRCK beta; MRCKb; myotonic dystrophy kinase related CDC42 binding kinase beta; myotonic dystrophy kinase-related CDC42-binding kinase beta; myotonic dystrophy protein kinase like beta; myotonic dystrophy protein kinase-like beta; serine/threonine-protein kinase MRCK beta |
| Gene Symbols | CDC42BPB |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?