missing translation for 'onlineSavingsMsg'
Learn More

MRCK Antibody, DyLight 594, Novus Biologicals™

Product Code. 18408055 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408055 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408055 Supplier Novus Biologicals Supplier No. NBP146823DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MRCK Polyclonal antibody specifically detects MRCK in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRCK
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias CDC42 binding protein kinase alpha (DMPK-like), CDC42 binidng protein kinase beta, CDC42-binding protein kinase alpha, CDC42-binding protein kinase alpha (DMPK-like), DMPK-like alpha, EC 2.7.11, EC 2.7.11.1, KIAA0451DKFZp686L1738, MRCK, MRCK alpha, MRCKADKFZp686P1738, Myotonic dystrophy kinase-related CDC42-binding kinase alpha, myotonic dystrophy kinase-related CDC42-binding protein kinase alpha, Myotonic dystrophy protein kinase-like alpha, PK428FLJ23347, serine/threonine-protein kinase MRCK alpha, ser-thr protein kinase PK428, ser-thr protein kinase related to the myotonic dystrophy protein kinase
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1610 and 1660 of human myotonic dystrophy protein kinase-like alpha using the numbering given in entry CAH71185.1 (GeneID 8476).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 8476
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.