missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRC2 Recombinant Rabbit Monoclonal Antibody (5I0I9)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557975

 View more versions of this product

Product Code. 30283423

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: DAALPEPNVF LIFSHGLQGC LEAQGGQVRV TPACNTSLPA QRWKWVSRNR LFNLGTMQCL GTGWPGTNTT ASLGMYECDR EALNLRWHCR TLGDQLSLLL GARTSNISKP GTLERGDQTR SGQWRIYGSE EDLCALPYHE VYTIQGNSHG KPCTIPFKYD NQWFHGCTST GREDGHLWCA TTQDYGKDER WGFCPIKSND CETFWDKDQL TDSCYQFNFQ STLSWREAWA SCEQQGADLL SITEIHEQTY INGLLTGYSS TLWIGLNDLD TSGGWQWSDN SPLKYLNWES DQPDNPSEEN CGVIRTESSG GWQNRDCSIA LPYVCKKKPN ATAEPTPPDR WANVKVECEP SWQPFQGHCY RLQAEKRSWQ ESKKACLRGG GDLVSIHSMA ELEFITKQIK QEVEELWIGL NDLKLQMNFE WSDGSLVSFT HWHPFEPNNF RDSLEDCVTI WGPEGRWNDS PCNQSLPSIC KKAGQLSQG.

May play a role as endocytotic lectin receptor displaying calcium-dependent lectin activity. Internalizes glycosylated ligands from the extracellular space for release in an endosomal compartment via clathrin-mediated endocytosis. May be involved in plasminogen activation system controlling the extracellular level of PLAUR/PLAU, and thus may regulate protease activity at the cell surface. May contribute to cellular uptake, remodeling and degradation of extracellular collagen matrices. May play a role during cancer progression as well as in other chronic tissue destructive diseases acting on collagen turnover. May participate in remodeling of extracellular matrix cooperating with the matrix metalloproteinases (MMPs).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MRC2
Recombinant Monoclonal
1.08 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q9UBG0
MRC2
Recombinant Protein (or fragment).
20 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
5I0I9
Unconjugated
MRC2
CD280; CLEC13E; collagen-binding factor Endo180; C-type lectin domain family 13 member E; C-type mannose receptor 2; Endo180; endocytic receptor (macrophage mannose receptor family); endocytic receptor 180; KIAA0709; Lectin lambda; macrophage mannose receptor 2; mannose receptor C type 2; mannose receptor, C type 2; mKIAA0709; Mrc2; novel lectin; UPARAP; UPAR-associated protein; urokinase-type plasminogen activator receptor-associated protein
Rabbit
Protein A
RUO
9902
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.