missing translation for 'onlineSavingsMsg'
Learn More

Mrc2 Antibody (2D7), Novus Biologicals™

Product Code. 18395609 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395609 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395609 Supplier Novus Biologicals Supplier No. H00009902M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mrc2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2D7
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006030
Gene Alias CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein
Host Species Mouse
Immunogen MRC2 (NP_006030, 105 a.a. ~ 214 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ASLGMYECDREALNLRWHCRTLGDQLSLLLGARTSNISKPGTLERGDQTRSGQWRIYGSEEDLCALPYHEVYTIQGNSHGKPCTIPFKYDNQWFHGCTSTGREDGHLWCA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline CD Markers, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 9902
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.