missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRAS Recombinant Rabbit Monoclonal Antibody (K01_2N36)
Description
MRAS Recombinant Monoclonal Antibody for Western Blot
MRAS is a member of the RAS superfamily of GTP-binding proteins which are membrane-anchored, intracellular signal transducers responsible for a variety of normal cellular functions. MRAS is oncogenically activated in a significant fraction of tumors.
Specifications
Specifications
| Antigen | MRAS |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2N36 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | MRAS |
| Gene Accession No. | O08989, O14807, P97538 |
| Gene Alias | 2900078C09Rik; AI326250; MRAS; M-RAs; Muscle and microspikes Ras; muscle RAS oncogene homolog; ras-related protein M-Ras; Ras-related protein R-Ras3; RRAS3; R-RAS3; Xras; X-Ras |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?