missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MRAP Polyclonal antibody specifically detects MRAP in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | MRAP |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | B27GCCD2, C21orf61chromosome 21 open reading frame 61, FALPFGD2, Fat cell-specific low molecular weight protein, Fat tissue-specific low MW protein, melanocortin 2 receptor accessory protein, melanocortin-2 receptor accessory protein |
| Host Species | Rabbit |
| Immunogen | melanocortin 2 receptor accessory protein. (Uniprot# Q8TCY5) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?