missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPZL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13158-1-AP-20UL
This item is not returnable.
View return policy
Description
Cell surface receptor, which is involved in signal transduction processes. Recruits PTPN11/SHP-2 to the cell membrane and is a putative substrate of PTPN11/SHP-2. Is a major receptor for concanavalin-A (ConA) and is involved in cellular signaling induced by ConA, which probably includes Src family tyrosine-protein kinases. Isoform 3 seems to have a domit negative role; it blocks tyrosine phosphorylation of MPZL1 induced by ConA. Isoform 1, but not isoform 2 and isoform 3, may be involved in regulation of integrin-mediated cell motility.Specifications
| MPZL1 | |
| Polyclonal | |
| Unconjugated | |
| MPZL1 | |
| MPZL1, myelin protein zero like 1, Protein zero related, PZR, PZR1b, PZRa, PZRb | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 360871, 68481, 9019 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95297, Q3TEW6, Q6AYT8 | |
| Mpzl1 | |
| MPZL1 Fusion Protein Ag3791 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction