missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPZL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566593
This item is not returnable.
View return policy
Description
Immunogen sequence: NSKRDYTGCST SESLSPVKQA PRKSPSDTEG LVKSLPSGSH QGPVIYAQLD HSGGHHSDKI NKSESVVYAD IRKN Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Cell surface receptor, which is involved in signal transduction processes. Recruits PTPN11/SHP-2 to the cell membrane and is a putative substrate of PTPN11/SHP-2. Is a major receptor for concanavalin-A (ConA) and is involved in cellular signaling induced by ConA, which probably includes Src family tyrosine-protein kinases. Isoform 3 seems to have a dominant negative role; it blocks tyrosine phosphorylation of MPZL1 induced by ConA. Isoform 1, but not isoform 2 and isoform 3, may be involved in regulation of integrin-mediated cell motility.
Specifications
| MPZL1 | |
| Polyclonal | |
| Unconjugated | |
| Mpzl1 | |
| 1110007A10Rik; FLJ21047; immunoglobulin family transmembrane protein; Mpzl1; MPZL1b; myelin protein zero like 1; myelin protein zero-like 1; myelin protein zero-like protein 1; protein zero related; protein zero-related; Pzr; PZR1b; PZRa; PZRb; UNQ849/PRO1787 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9019 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95297 | |
| Mpzl1 | |
| Recombinant Human MPZL1. Recombinant protein control fragment (Product #RP-107247). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction