Learn More
CPC (Colder) MPX™ Quick-Disconnect Fittings, Hose Barb Body, Non-Valved
Product Code. 30053034
Click to view available options
Color:
Amber tint
Purple Tint
Hose Barb:
Hose Barb Body
Hose Barb Body with Lock
Unit Size:
Pack of 25
Product Code. 30053034
Brand: CPC (Colder) 5011334
Only null left
Add to basket
Add to basket
Connect to a coupling insert to quickly create a sterile fluid pathway
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Connect to a male coupling inserts for easy media transfer. Operate ergonomic thumb latch even with gloved hands. Mix and match termination sizes for added flexibility. 0.5 in. nominal flow path size
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/2 in.
Material Polycarbonate
Min. Operating Temperature (Metric) -40°F
Packaging Quantity 25/Pk.
Hose Barb Hose Barb Body with Lock
Outlet Port Quick Disconnect Body x Hose Barb
Shape Straight
Color Purple Tint