missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPV17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112612
This item is not returnable.
View return policy
Description
MPV17 Polyclonal Antibody for Western Blot, ELISA
This gene encodes a mitochondrial inner membrane protein that is implicated in the metabolism of reactive oxygen species. Mutations in this gene have been associated with the hepatocerebral form of mitochondrial DNA depletion syndrome.
Specifications
| MPV17 | |
| Polyclonal | |
| Unconjugated | |
| MPV17 | |
| hMPV; h-MPV; Mpv17; Mpv-17; MpV17 mitochondrial inner membrane protein; MPV17 mitochondrial membrane protein-like; Mpv17 protein; Mpv17 transgene, kidney disease mutant-like; Mpv17, human homolog of glomerulosclerosis and nephrotic syndrome; MPV17, mitochondrial inner membrane protein; Mpv17l; MTDPS6; OTTHUMP00000122591; Protein Mpv17; SYM1; Tg.Mpv17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17527, 4358 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P19258, P39210 | |
| MPV17 | |
| Recombinant Human Protein Mpv17 protein (1-176AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur