missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MPure™ Viral RNA Extraction Kit
Shop All MP Biomedicals Products
Click to view available options
:
48 preps
Description
- Completely automated purification protocol
- Run times as little as 40 minutes
- RNA can be used immediately in downstream applications
Specifications
Specifications
| Content And Storage | Store at room temperature |
| Description | Viral RNA Extraction Kit |
| Isolation Technology | Magnetic Bead |
| Sample Type | Human Biological Specimens (Plasma, Serum, Other Cell-free Fluids) |
| For Use With (Equipment) | MPure-12 Automated Nucleic Acid Purification System |
| For Use With (Application) | RNA Purification |
| Includes | Proteinase K, 30μL; Lysis Buffer 2, 720μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL; RNase Free water, 1000μL; RNase Free water, 1000μL |
| No. of Reactions | 48 Preps |
| Sample Size | 100 to 400μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction