missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MPure™ Tissue DNA Extraction Kit
Shop All MP Biomedicals Products
Click to view available options
:
48 preps
Description
MPure Tissue DNA Extraction Kit contains everything needed for sample preparation. However, for tough tissues such as cardiac or skeletal muscle, spleen, intestinal or other connective tissue-rich samples, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time.
- Completely automated purification protocol
- Runs in as little as 40 minutes
- Produces nucleic acids that can be used immediately in downstream applications
Specifications
Specifications
| Content And Storage | Store at room temperature |
| Description | Tissue DNA Extraction Kit |
| Isolation Technology | Magnetic Bead |
| Sample Type | Animal Tissue, Blood Stain, Swab |
| For Use With (Equipment) | MPure-12 Automated Nucleic Acid Purification System |
| For Use With (Application) | Tissue DNA Purification |
| Includes | Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer, 1000μL; Elution Buffer, 1000μL |
| No. of Reactions | 48 Preps |
| Sample Volume | Purify DNA from 100 - 400μL or 10 - 40mg of tissue; 100 - 400μL or one swab or brush for dried swab samples; 100 - 400μL or four discs for dried blood |
| Test Time | As little as 40 minutes |
Safety and Handling
Product Identifier
- MPure Tissue DNA Extraction Kit
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contains : proteinase, sodium dodecyl sulphate, ethanol, guanidium chloride, polyethylene glycol, propan-2-ol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction