missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Plant DNA Extraction Kit

For extraction of genomic DNA from plant and fungal tissues

Brand:  MP Biomedicals™ 117022150

Additional Details : Weight : 0.30000kg Transport : UN number : 3316 Chem class : 9 Pack group : II


Product Code. 15709210

  • £319.06 / 1 set
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL

Description

Description

MPure Plant DNA Extraction Kit contains everything needed for sample preparation. However, for tough tissues such as roots and seeds, or tough fungal tissue, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can increase yields as well as shorten processing time.

  • Compatible with fresh or lyophilized plant tissue as well as fully formed mycelium or cells from yeast culture
  • Completely automated purification protocol
  • Runs in as little as 40 minutes
  • Generates DNA that can be used immediately in downstream applications

Specifications

Specifications

Store at room temperature
Magnetic Bead
DNA Purification
48 Preps
Plant DNA Extraction Kit
MPure-12 Automated Nucleic Acid Purification System
Lysis Buffer 2, 720μL; Binding Buffer 1, 720μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
As little as 40 minutes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P101-If medical advice is needed, have product container or label at hand.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash hands thoroughly after handling.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P270-Do not eat, drink or smoke when using this product.
  • P240-Ground/bond container and receiving equipment.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P270-Do not eat, drink or smoke when using this product.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P240-Ground/bond container and receiving equipment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P242-Use only non-sparking tools.
  • P233-Keep container tightly closed.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P242-Use only non-sparking tools.
  • P271-Use only outdoors or in a well-ventilated area.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P243-Take precautionary measures against static discharge.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P370+P378-In case of fire: Use to extinguish.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P271-Use only outdoors or in a well-ventilated area.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
  • P370+P378-In case of fire: Use to extinguish.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.

Supplemental information
  • MIXTURE LIST-Contains : éthanol, guanidine hydrochloride, triton

SDS
Documents

Documents

Certificates
Special Offers

Special Offers