missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Forensic DNA Extraction Kit

Product Code. 15769200 Shop All MP Biomedicals Products
Change view
Click to view available options
No. of Reactions:
48 Preps
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
15769200 48 Preps 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15769200 Supplier MP Biomedicals™ Supplier No. 117022110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extract and isolate genomic DNA from forensic samples

The kit contains everything needed for sample preparation. However, for samples such as swipes and swabs, cigarette butts and bodily fluid stained fabrics or other solid matrices, sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing time. DNA extracted using the MPure Forensic DNA Extraction Kit is ready for immediate use in downstream applications.

  • Completely automated purification protocol
  • Run in as little as 40 minutes
  • DNA is ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Forensic DNA Extraction Kit
Sample Type Forensic Sample
For Use With (Equipment) MPure-12™ Automated Nucleic Acid Purification System
For Use With (Application) Nucleic Acid Purification; Genomic DNA from Forensic samples
Includes Lysis Buffer 2, 1000μL; Binding Buffer 1, 1000μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Sample Volume Clotted/ dried blood: 20μL; Forensic Surface and Contact Swabs, Hair root: 0.5 to 1cm; Human tissues: 40mg; Saliva: 50μL; Sperm Stains: 5 to 10μL or 1cm2 of the forensic sample; Chewing Gum: 40 mg; Cigarette butts: 1cm2 paper from the end of the cigarette or filter
Test Time As little as 40 minutes
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • MPure Forensic DNA Extraction Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: urea, sodium dodecylsulphate, ethanol,guanidine hydrochloride, Triton X-100, isopropanol, glycerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.