missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ MPure™ Blood DNA Extraction Kit 200

Product Code. 15759200 Shop All MP Biomedicals Products
Click to view available options
:
48 preps
This item is not returnable. View return policy

Product Code. 15759200

Brand: MP Biomedicals™ 117022100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For extraction of DNA from mammalian whole blood or a suspension of mammalian blood cells

MPure Blood DNA Extraction Kit 200 is used with the MPure-12 aNAP System for extraction of DNA from 100-400μL mammalian whole blood or suspension of mammalian blood cells. Nucleic acids extracted from MPure Blood DNA Extraction Kit 200 can be used in a number of downstream application including: PCR, qPCR, Sequencing (NGS), Microarray, RFLP, and Southern Blot Analysis.

  • For extraction of host gDNA, mitochondrial and viral DNA
  • Extracts DNA from 100-400µL mammalian whole blood
  • Nucleic acids can be used immediately in downstream applications
  • Completely automated
  • Can be run in as little as 50 minutes

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature
Description Blood DNA Extraction Kit 200
Sample Type Buffy Coat, Leukocytes, Whole Blood Cells
For Use With (Equipment) MPure-12 Automated Nucleic Acid Purification System
For Use With (Application) Nucleic Acid Purification
Includes Proteinase K solution, 40μL; Lysis Buffer 2, 1000μL; Binding Buffer 1, 600μL; Magnetic Bead Solution, 800μL; Washing Buffer 1, 1000μL; Washing Buffer 2, 1000μL; Washing Buffer 3, 1000μL; Elution Buffer 1, 1000μL; Elution Buffer 2, 1000μL
No. of Reactions 48 Preps
Sample Volume 100-400μL whole blood (WBC count less than 2 x 104 cells/μL); 100-400μL leukocyte concentration (contain no more than 5 x 106 cells) ; 100-400μL buffy coat
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • MPure Blood DNA Extraction Kit 200
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethyl alcohol, guanidine hydrochloride,triton x-100, isopropanol, glycerol, proteinase K
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.