missing translation for 'onlineSavingsMsg'
Learn More

MPTP hydrochloride, MedChemExpress

Product Code. 16426836
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16426836

Brand: Medchem Express HY15608

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MPTP hydrochloride is a brain penetrant dopamine neurotoxin, inducing Parkinson’s Disease. MPTP hydrochloride, a precusor of MPP+, induces apoptosis.

MPTP hydrochloride is a brain penetrant dopamine neurotoxin, inducing Parkinson’s Disease. MPTP hydrochloride, a precusor of MPP+, induces apoptosis.

Specifications

Chemical Name or Material MPTP hydrochloride
Physical Form Solid
CAS 23007-85-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H16ClN
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 12 mg/mL (57.22 mM; Need ultrasonic and warming) ∣H2O : 10 mg/mL (47.68 mM; Need ultrasonic)
SMILES CN1CC=C(C2=CC=CC=C2)CC1.[H]Cl
Molecular Weight (g/mol) 209.72
Formula Weight 209.72
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.