missing translation for 'onlineSavingsMsg'
Learn More

MPST Antibody (1B5), Novus Biologicals™

Product Code. 18392378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392378 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392378 Supplier Novus Biologicals Supplier No. H00004357M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MPST Monoclonal antibody specifically detects MPST in Human samples. It is validated for ELISA, Immunoprecipitation, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPST
Applications ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 1B5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001013454
Gene Alias mercaptopyruvate sulfurtransferase, MGC24539, MSThuman liver rhodanese, TST2EC 2.8.1.2,3-mercaptopyruvate sulfurtransferase
Host Species Mouse
Immunogen MPST (NP_001013454.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MASPQLCRALVSAQWVAEALRAPRAGQPLQLLDASWYLPKLGRDARREFEERHIPGAAFFDIDQCSDRTSPYDHMLPGAEHFAEYAGRLGVGAATHVVIY
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4357
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.