Learn More
Invitrogen™ MPS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579936
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Caco-2 whole cell, human RT4 whole cell, human Jurkat whole cell, rat PC-12 whole cell, rat RH35 whole cell, mouse RAW2647 whole cell, mouse SP2/0 whole cell. IHC: human intestinal cancer tissue, mouse brain cancer tissue, rat brain cancer tissue. ICC/IF: U20S cell. Flow: HeLa cell.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S27E family of ribosomal proteins. It contains a C4-type zinc finger domain that can bind to zinc. The encoded protein has been shown to be able to bind to nucleic acid. It is located in the cytoplasm as a ribosomal component, but it has also been detected in the nucleus. Studies in rat indicate that ribosomal protein S27 is located near ribosomal protein S18 in the 40S subunit and is covalently linked to translation initiation factor eIF3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| MPS1 | |
| Polyclonal | |
| Unconjugated | |
| RPS27 | |
| 3200001M24Rik; 40S ribosomal protein S27; metallopanstimulin 1; Metallopan-stimulin 1; MPS1; MPS-1; RGD:621045}; ribosomal protein S27; RP11-422P24.3; rps27; rps27 {ECO:0000312; S27; S27-1; Small ribosomal subunit protein eS27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57294, 6232, 94266 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P42677, Q6ZWU9, Q71TY3 | |
| RPS27 | |
| E. coli-derived human MPS1 recombinant protein (Position: P2-H84). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.