missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550186
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MPS1 protein.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S27E family of ribosomal proteins. It contains a C4-type zinc finger domain that can bind to zinc. The encoded protein has been shown to be able to bind to nucleic acid. It is located in the cytoplasm as a ribosomal component, but it has also been detected in the nucleus. Studies in rat indicate that ribosomal protein S27 is located near ribosomal protein S18 in the 40S subunit and is covalently linked to translation initiation factor eIF3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| MPS1 | |
| Polyclonal | |
| Unconjugated | |
| RPS27 | |
| 3200001M24Rik; 40S ribosomal protein S27; metallopanstimulin 1; Metallopan-stimulin 1; MPS1; MPS-1; RGD:621045}; ribosomal protein S27; RP11-422P24.3; rps27; rps27 {ECO:0000312; S27; S27-1; Small ribosomal subunit protein eS27 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57294, 6232, 94266 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P42677, Q6ZWU9, Q71TY3 | |
| RPS27 | |
| Fusion protein corresponding to residues near the N terminal of human ribosomal protein S27. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction