missing translation for 'onlineSavingsMsg'
Learn More

Mps1-IN-3, MedChemExpress

Product Code. 16435856
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16435856

Brand: Medchem Express HY12401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mps1-IN-3 is a potent and selective MPS1 kinase inhibitor, with an IC50 of 50 nM.

Mps1-IN-3 is a potent and selective MPS1 kinase inhibitor, with an IC50 of 50 nM.

Specifications

Chemical Name or Material Mps1-IN-3
CAS 1609584-72-6
Color Light Green
Purity Grade Notes Research
Molecular Formula C26H31N7O4S
Quantity 100 mg
Solubility Information DMSO : 31.25 mg/mL (58.13 mM; ultrasonic and warming and heat to 60°C)
SMILES OC1CCN(C2=CC=C(NC3=NC(NC4=CC=CC=C4S(=O)(C(C)C)=O)=C5N=CNC5=N3)C(OC)=C2)CC1
Molecular Weight (g/mol) 537.63
Formula Weight 537.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.