missing translation for 'onlineSavingsMsg'
Learn More

Mps1-IN-1, MedChemExpress

Product Code. 16485628
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485628 10 mg 10mg
16461797 50 mg 50mg
16438327 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485628 Supplier Medchem Express Supplier No. HY13298

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mps1-IN-1 is a potent, selective and ATP-competitive Mps1 kinase inhibitor, with an IC50 and a Kd of 367 nM and 27 nM.

Mps1-IN-1 is a potent, selective and ATP-competitive Mps1 kinase inhibitor, with an IC50 and a Kd of 367 nM and 27 nM.

Specifications

Chemical Name or Material Mps1-IN-1
CAS 1125593-20-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H33N5O4S
Quantity 10 mg
Solubility Information DMSO : ≥ 39 mg/mL (72.81 mM)
SMILES O=S(C(C=CC=C1)=C1NC2=CC(NC3=C(OC)C=C(N4CCC(O)CC4)C=C3)=NC5=C2C=CN5)(C(C)C)=O
Molecular Weight (g/mol) 535.66
Formula Weight 535.66
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.