missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPRIP Polyclonal Antibody

MPRIP Polyclonal Antibody
Brand: Thermo Scientific PA554559
This item is not returnable.
View return policy
Description
MPRIP Polyclonal Antibody for IHC
Specifications
| MPRIP | |
| Polyclonal | |
| Unconjugated | |
| MPRIP | |
| 9530046C02; AA536749; AI647711; C76423; KIAA0864; mKIAA0864; Mprip; Mrip; M-RIP; myosin phosphatase Rho interacting protein; myosin phosphatase Rho-interacting protein; myosin phosphatase-Rho interacting protein; p116 Rho interacting protein; p116Rip; Rho interacting protein 3; rho-interacting protein 3; Rho-interacting protein 3 (p116RIP) (RIP3); RHOIP3; RIP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23164 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6WCQ1 | |
| MPRIP | |
| Recombinant protein corresponding to Human MPRIP. Recombinant protein control fragment (Product #RP-93317). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction