missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPRIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£225.00 - £451.00
Specifications
| Antigen | MPRIP |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18460181
|
Novus Biologicals
NBP1-81035-25ul |
25ul |
£225.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18756093
|
Novus Biologicals
NBP1-81035 |
0.1 mL |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MPRIP Polyclonal specifically detects MPRIP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MPRIP | |
| Polyclonal | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q6WCQ1 | |
| 23164 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KDAYELEVLLRVKESEIQYLKQEISSLKDELQTALRDKKYASDKYKDIYTELSIAKAKADCDISRLKEQLKAATEALGEKSPDSATVSGYDIMKSKSNPDFLKKDRSCVTRQL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| KIAA0864Rho interacting protein 3, MRIPP116RIP, myosin phosphatase Rho interacting protein, p116Ripmyosin phosphatase-Rho interacting protein, RHOIP3myosin phosphatase Rho-interacting protein, RIP3M-RIPRho-interacting protein 3 | |
| MPRIP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title